2-(1H-Pyrazol-1-yl)benzaldehyde 95%
Supplier: Apollo Scientific
Warning
OR1649-1G OR1649-5G
APOSOR1649-1GEA 72.5 EUR
APOSOR1649-1G APOSOR1649-5G
2-(1H-Pyrazol-1-yl)benzaldehyde 95%
2-(1H-Pyrazol-1-yl)benzaldehyde
Formula: C₁₀H₈N₂O
Melting Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD03086186
CAS Number: 138479-47-7