1,5-Dibromonaphthalene 98%
Supplier: Apollo Scientific
Warning
OR16492-1G OR16492-5G
APOSOR16492-1GEA 29 EUR
APOSOR16492-1G APOSOR16492-5G
1,5-Dibromonaphthalene 98%
1,5-Dibromonaphthalene
Formula: C₁₀H₆Br₂
MW: 285.9600 g/mol
Melting Pt: 130…130.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00089984
CAS Number: 7351-74-8