Benzylamine 98%
Supplier: Apollo Scientific
Danger
Synonyms: α-Aminotoluene
OR16591-1KG OR16591-250G OR16591-5KG
APOSOR16591-1KGEA 87 EUR
APOSOR16591-1KG APOSOR16591-250G APOSOR16591-5KG
Benzylamine 98%
Benzylamine
Formula: (C₆H₅)CH₂NH₂
Boiling Pt: 183…185 °C
Melting Pt: 10 °C
Density: 0.981 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8,III