(3-Morpholinophenyl)methanol 97%
Supplier: Apollo Scientific
Warning
Synonyms: (3-morpholin-4-ylphenyl)methanol, Benzenemethanol,3-(4-morpholinyl)-, 3-(Morpholin-4-yl)benzyl alcohol, 3-Morpholinobenzyl Alcohol
OR16605-1G OR16605-250MG
APOSOR16605-1GEA 140 EUR
APOSOR16605-1G APOSOR16605-250MG
(3-Morpholinophenyl)methanol 97%
(3-Morpholinophenyl)methanol
Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Melting Pt: 51…54 °C
Storage Temperature: Ambient
MDL Number: MFCD07772812
CAS Number: 145127-38-4