6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one 97%
Supplier: Apollo Scientific
Warning
OR16671-1G OR16671-5G OR16671-10G
APOSOR16671-1GEA 21.9 EUR
APOSOR16671-1G APOSOR16671-5G APOSOR16671-10G
6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one 97%
6-Bromo-3,4-dihydro-1H-[1,8]naphthyridin-2-one
Formula: C₈H₇BrN₂O
MW: 227.0600 g/mol
Melting Pt: 267…270 °C
Storage Temperature: Ambient
MDL Number: MFCD05664027
CAS Number: 129686-16-4