tert-Butyl-7-benzyl-2,7-diazaspiro[3.5]nonane-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 7-benzyl-2,7-diazaspiro[3.5]nonane-2-carboxylate, 7-Benzyl-2,7-diazaspiro[3.5]nonane, N2-BOC protected
OR16700-100MG OR16700-1G OR16700-250MG
APOSOR16700-100MGEA 98.6 EUR
APOSOR16700-100MG APOSOR16700-1G APOSOR16700-250MG
tert-Butyl-7-benzyl-2,7-diazaspiro[3.5]nonane-2-carboxylate
tert-Butyl-7-benzyl-2,7-diazaspiro[3.5]nonane-2-carboxylate
Formula: C₁₉H₂₈N₂O₂
MW: 316.4400 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111843
CAS Number: 929301-99-5