8-Boc-2,8-diazaspiro[4.5]decane oxalate
Supplier: Apollo Scientific
Warning
Synonyms: 2,8-Diazaspiro[4.5]decane oxalate, N8-BOC protected
OR16707-1G OR16707-250MG
APOSOR16707-1GEA 429 EUR
APOSOR16707-1G APOSOR16707-250MG
8-Boc-2,8-diazaspiro[4.5]decane oxalate
8-Boc-2,8-diazaspiro[4.5]decane oxalate
Formula: C₁₅H₂₆N₂O₆
Storage Temperature: Ambient
MDL Number: MFCD20527269
CAS Number: 1408074-53-2