Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-Chloro-5-azaindole-2-carboxylate
OR16875-1G OR16875-250MG
APOSOR16875-1GEA 133 EUR
APOSOR16875-1G APOSOR16875-250MG
Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate
Methyl-4-chloro-1H-pyrrolo[3,2-c]pyridine-2-carboxylate
Formula: C₉H₇ClN₂O₂
MW: 210.62 g/mol
Melting Pt: 224…225 °C
Storage Temperature: Ambient
MDL Number: MFCD09870065
CAS Number: 688357-19-9