Iodoethane 98%
Supplier: Apollo Scientific
Danger
Synonyms: Ethyl iodide
OR16933-100G OR16933-500G
APOSOR16933-100GEA 87 EUR
APOSOR16933-100G APOSOR16933-500G
Iodoethane 98%
Iodoethane
Formula: C₂H₅I
Boiling Pt: 69…73 °C
Melting Pt: –108 °C
Density: 1.95 g/cm³ (25 °C)
Flash Pt: 53 °C
Storage Temperature: Ambient
MDL Number: MFCD00001091
CAS Number: 75-03-6
EINECS: 200-833-1
UN: 1993
ADR: 3,III