2,6-Dibromobenzenecarbaldehyde 95%
Supplier: Apollo Scientific
Warning
Synonyms: 2,6-Dibromobenzaldehyde
OR16948-1G OR16948-5G
APOSOR16948-1GEA 21.9 EUR
APOSOR16948-1G APOSOR16948-5G
2,6-Dibromobenzenecarbaldehyde 95%
2,6-Dibromobenzenecarbaldehyde
Formula: C₇H₄Br₂O
MW: 263.93 g/mol
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD11040410
CAS Number: 67713-23-9