1-(3-Chloropropyl)-4-methylpiperazine
Supplier: Apollo Scientific
Warning
OR16966-1G OR16966-5G
APOSOR16966-1GEA 0 EUR
APOSOR16966-1G APOSOR16966-5G
1-(3-Chloropropyl)-4-methylpiperazine
1-(3-Chloropropyl)-4-methylpiperazine
Formula: C₈H₁₇ClN₂
Boiling Pt: 85…87 °C/7mmHg
Melting Pt: –23…–22 °C
Density: 1.015 g/cm³ (25 °C)
Flash Pt: 101 °C
Storage Temperature: Ambient
CAS Number: 104-16-5
EINECS: 203-181-6