3,5-Di-tert-butylbenzaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 3,5-Bis(tert-butyl)benzaldehyde
OR16972-1G OR16972-25G OR16972-5G
APOSOR16972-1GEA 21.9 EUR
APOSOR16972-1G APOSOR16972-25G APOSOR16972-5G
3,5-Di-tert-butylbenzaldehyde
3,5-Di-tert-butylbenzaldehyde
Formula: C₁₅H₂₂O
MW: 218.34 g/mol
Boiling Pt: 130…132 °C/5mmHg
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00026298
CAS Number: 17610-00-3