3-Amino-o-cresol
Supplier: Apollo Scientific
Warning
Synonyms: 3-Amino-2-methylphenol, 1-Amino-3-hydroxy-2-methylbenzene, 3-Hydroxy-2-methylaniline, 2-Amino-6-hydroxytoluene
OR16980-25G OR16980-5G
APOSOR16980-25GEA 395 EUR
APOSOR16980-25G APOSOR16980-5G
3-Amino-o-cresol
3-Amino-o-cresol
Formula: C₇H₉NO
MW: 123.15 g/mol
Melting Pt: 129…130 °C
Storage Temperature: Ambient
MDL Number: MFCD00007787
CAS Number: 53222-92-7