3-(3-Chlorophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Danger
OR17-1G OR17-250MG
APOSOR17-1GEA 139 EUR
APOSOR17-1G APOSOR17-250MG
3-(3-Chlorophenyl)-1H-pyrazole
3-(3-Chlorophenyl)-1H-pyrazole
Formula: C₉H₇ClN₂
Storage Temperature: Ambient
MDL Number: MFCD09860599
CAS Number: 59843-69-5