trans-N1,N2-Dimethylcyclohexane-1,2-diamine
Supplier: Apollo Scientific
Danger
Synonyms: trans-N,N'-Dimethyl-1,2-cyclohexanediamine
OR17015-1G OR17015-25G OR17015-5G
APOSOR17015-1GEA 21.9 EUR
APOSOR17015-1G APOSOR17015-25G APOSOR17015-5G
trans-N1,N2-Dimethylcyclohexane-1,2-diamine
trans-N1,N2-Dimethylcyclohexane-1,2-diamine
Formula: C₈H₁₈N₂
Boiling Pt: 78…80 °C/18mmHg
Density: 0.902 g/cm³ (25 °C)
Flash Pt: 74 °C
Storage Temperature: Ambient
MDL Number: MFCD03001702
CAS Number: 67579-81-1
UN: 2735
ADR: 8,II