3-Bromo-p-cresol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Bromo-4-methylphenol, 1-Bromo-3-hydroxy-6-methylbenzene
OR17017-1G OR17017-25G OR17017-5G
APOSOR17017-1GEA 21.9 EUR
APOSOR17017-1G APOSOR17017-25G APOSOR17017-5G
3-Bromo-p-cresol 98%
3-Bromo-p-cresol
Formula: C₇H₇BrO
MW: 187.04 g/mol
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD08273793
CAS Number: 60710-39-6