Methyl-2-oxo-2H-pyran-3-carboxylate 98%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 2-oxo-2H-pyran-3-carboxylate
OR17032-1G OR17032-5G OR17032-25G
APOSOR17032-1GEA 21.9 EUR
APOSOR17032-1G APOSOR17032-5G APOSOR17032-25G
Methyl-2-oxo-2H-pyran-3-carboxylate 98%
Methyl-2-oxo-2H-pyran-3-carboxylate
Formula: C₇H₆O₄
MW: 154.12 g/mol
Boiling Pt: 146…148 °C/0.75mmHg
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00075443
CAS Number: 25991-27-9