2,4-Dibromomesitylene
Supplier: Apollo Scientific
|
Warning
|
OR17043-25G
OR17043-5G
APOSOR17043-25GEA
31.9
EUR
APOSOR17043-25G
APOSOR17043-5G
2,4-Dibromomesitylene
2,4-Dibromomesitylene
Formula:
C₉H₁₀Br₂ Boiling Pt: 278…279 °C Melting Pt: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013525 CAS Number: 6942-99-0 EINECS: 230-100-1 |