1-Iodonaphthalene 97+%
Supplier: Apollo Scientific
|
Warning
|
OR17044-100G
OR17044-25G
OR17044-5G
APOSOR17044-100GEA
165
EUR
APOSOR17044-100G
APOSOR17044-25G
APOSOR17044-5G
1-Iodonaphthalene 97+%
1-Iodonaphthalene
Formula:
C₁₀H₇I Boiling Pt: 302…305 °C Melting Pt: 44443 °C Density: 1.74 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003876 CAS Number: 90-14-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...