Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) 97%
Supplier: Apollo Scientific
Warning
Synonyms: (A-taPhos)2PdCl2, PdCl2(Amphos)2, Bis(di-tert-butyl(4-dimethylaminophenyl)phosphine)dichloropalladium(II), Bis[di-tert-butyl(4-dimethylaminophenyl)phosphine]dichloropalladium, Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
OR17048-10G OR17048-1G OR17048-5G
APOSOR17048-10GEA 542 EUR
APOSOR17048-10G APOSOR17048-1G APOSOR17048-5G
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II) 97%
Dichlorobis[di-tert-butyl(4-dimethylaminophenyl)phosphine]palladium(II)
Formula: C₃₂H₅₆Cl₂N₂P₂Pd
MW: 708.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265123
CAS Number: 887919-35-9