4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester
Supplier: Apollo Scientific
Warning
Synonyms: 4-Amino-1H-indazole, N1-BOC protected, tert-Butyl 4-amino-1H-indazole-1-carboxylate
OR17050-1G OR17050-250MG
APOSOR17050-1GEA 69.6 EUR
APOSOR17050-1G APOSOR17050-250MG
4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester
4-Amino-1H-indazole-1-carboxylic acid tert-butyl ester
Formula: C₁₂H₁₅N₃O₂
MW: 233.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11052618
CAS Number: 801315-74-2