p-Toluonitrile 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Methylbenzonitrile, p-Methylbenzonitrile
OR1709-1KG OR1709-250G OR1709-50G
APOSOR1709-1KGEA 377 EUR
APOSOR1709-1KG APOSOR1709-250G APOSOR1709-50G
p-Toluonitrile 97%
p-Toluonitrile
Formula: C₈H₇N
Boiling Pt: 217 °C
Melting Pt: 26…28 °C
Density: 0.981 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00001827
CAS Number: 104-85-8