2-Bromo-5-iodonitrobenzene 97%
Supplier: Apollo Scientific
Warning
Synonyms: 1-Bromo-4-Iodo-2-nitrobenzene
OR17118-1G OR17118-250MG OR17118-5G
APOSOR17118-1GEA 37.7 EUR
APOSOR17118-1G APOSOR17118-250MG APOSOR17118-5G
2-Bromo-5-iodonitrobenzene 97%
2-Bromo-5-iodonitrobenzene
Formula: C₆H₃BrINO₂
MW: 327.9000 g/mol
Melting Pt: 93…95 °C
Storage Temperature: Ambient
MDL Number: MFCD12547765
CAS Number: 713512-18-6