1,4-Dibromobenzene 99%
Supplier: Apollo Scientific
Warning
OR17127-1KG OR17127-250G OR17127-2KG
APOSOR17127-1KGEA 33.5 EUR
APOSOR17127-1KG APOSOR17127-250G APOSOR17127-2KG
1,4-Dibromobenzene 99%
1,4-Dibromobenzene
Formula: C₆H₄Br₂
MW: 235.91 g/mol
Boiling Pt: 219…220 °C
Melting Pt: 83…90 °C
Density: 1.841 g/cm³ (25 °C)
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00000089
CAS Number: 106-37-6
EINECS: 203-390-2
UN: 3077
ADR: 9+EHS,III