Acetonedimethyl acetal 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane
OR17208-2.5L OR17208-500ML
APOSOR17208-2.5LEA 207 EUR
APOSOR17208-2.5L APOSOR17208-500ML
Acetonedimethyl acetal 98%
Acetone-dimethyl acetal
Formula: C₅H₁₂O₂
Boiling Pt: 79…81 °C
Melting Pt: –47 °C
Density: 0.848 g/cm³ (25 °C)
Flash Pt: –11 °C
Storage Temperature: Ambient
MDL Number: MFCD00008479
CAS Number: 77-76-9
EINECS: 201-056-0
UN: 3271
ADR: 3,II