Methyl 4-Bromo-2-chlorobenzoate 97%
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 4-bromo-2-chlorobenzoate, 4-Bromo-2-chlorobenzoic acid methyl ester
OR17252-10G OR17252-5G OR17252-1G
APOSOR17252-10GEA 98.6 EUR
APOSOR17252-10G APOSOR17252-5G APOSOR17252-1G
Methyl 4-Bromo-2-chlorobenzoate 97%
Methyl 4-Bromo-2-chlorobenzoate
Formula: C₈H₆BrClO₂
MW: 249.49 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10566823
CAS Number: 185312-82-7