4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine
Supplier: Apollo Scientific
Danger
Synonyms: 6-Amino-4-chloro-1H-pyrazolo[3,4-d]pyrimidine
OR17302-5G
APOSOR17302-5GEA 143 EUR
APOSOR17302-5G
4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine
4-Chloro-1H-pyrazolo[3,4-d]pyrimidin-6-amine
Formula: C₅H₄ClN₅
MW: 169.5700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08059805
CAS Number: 100644-65-3
UN: 2811
ADR: 6.1,III