1-Boc-piperazine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
tert-Butyl 1-piperazinecarboxylate, tert-butyl piperazine-1-carboxylate
OR1731-100G
OR1731-250G
APOSOR1731-100GEA
26.1
EUR
APOSOR1731-100G
APOSOR1731-250G
1-Boc-piperazine 98%
1-Boc-piperazine
Formula:
C₉H₁₈N₂O₂ Boiling Pt: 98…100 °C Melting Pt: 43…48 °C Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075265 CAS Number: 57260-71-6 |