Ethyl-2-(bromomethyl)acrylate 95%
Supplier: Apollo Scientific
Danger
Synonyms: Ethyl 2-(bromomethyl)acrylate
OR17520-5G OR17520-25G
APOSOR17520-5GEA 21.9 EUR
APOSOR17520-5G APOSOR17520-25G
Ethyl-2-(bromomethyl)acrylate 95%
Ethyl-2-(bromomethyl)acrylate
Formula: C₆H₉BrO₂
MW: 193.04 g/mol
Boiling Pt: 38 °C/0.8mmHg
Density: 1.398 g/cm³ (25 °C)
Flash Pt: 81 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00031518
CAS Number: 17435-72-2
UN: 3265
ADR: 8,III