2,6-Dichloro-3-methylpyridine 98%
Supplier: Apollo Scientific
Warning
OR17532-25G
APOSOR17532-25GEA 81.2 EUR
APOSOR17532-25G
2,6-Dichloro-3-methylpyridine 98%
2,6-Dichloro-3-methylpyridine
Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Melting Pt: 54…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00137847
CAS Number: 58584-94-4