3-Bromo-2-methyl-1-propene
Supplier: Apollo Scientific
Danger
Synonyms: 3-Bromo-2-methylpropene, 2-Methyl-2-propenyl Bromide, 2-Methylallyl Bromide
OR17610-100G OR17610-25G OR17610-5G
APOSOR17610-100GEA 284 EUR
APOSOR17610-100G APOSOR17610-25G APOSOR17610-5G
3-Bromo-2-methyl-1-propene
3-Bromo-2-methyl-1-propene
Formula: C₄H₇Br
MW: 135.00 g/mol
Boiling Pt: 94…95 °C
Density: 1.339 g/cm³ (25 °C)
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134155
CAS Number: 1458-98-6
UN: 2924
ADR: 3+8,II