4-Bromo-2,6-dinitrotoluene
Supplier: Apollo Scientific
Warning
OR17614-1G OR17614-5G
APOSOR17614-1GEA 36.4 EUR
APOSOR17614-1G APOSOR17614-5G
4-Bromo-2,6-dinitrotoluene
4-Bromo-2,6-dinitrotoluene
Formula: C₇H₅BrN₂O₄
MW: 261.03 g/mol
Melting Pt: 86…89 °C
Storage Temperature: Ambient
MDL Number: MFCD07357280
CAS Number: 95192-64-6
UN: 2811
ADR: 6.1,III