4-Bromo-α-methylbenzyl alcohol 98%
Supplier: Apollo Scientific
Synonyms: 4-Bromophenyl methyl carbinol, 1-(4-Bromophenyl)ethanol, p-Bromo-α-methylbenzyl alcohol
OR17616-100G OR17616-25G OR17616-500G
APOSOR17616-100GEA 198 EUR
APOSOR17616-100G APOSOR17616-25G APOSOR17616-500G
4-Bromo-α-methylbenzyl alcohol 98%
4-Bromo-α-methylbenzyl alcohol
Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 119…121 °C/7mmHg
Melting Pt: 36…38 °C
Density: 1.463 g/cm³ (25 °C)
Flash Pt: 63 °C
Storage Temperature: Ambient
MDL Number: MFCD00004514
CAS Number: 5391-88-8