3-Amino-6-bromoisoquinoline 97%
Supplier: Apollo Scientific
Warning
Synonyms: 6-Bromoisoquinolin-3-amine
OR17655-1G OR17655-5G
APOSOR17655-1GEA 78.3 EUR
APOSOR17655-1G APOSOR17655-5G
3-Amino-6-bromoisoquinoline 97%
3-Amino-6-bromoisoquinoline
Formula: C₉H₇BrN₂
MW: 223.0700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11101002
CAS Number: 891785-28-7