5-Chloro-o-toluidine 98%
Supplier: Apollo Scientific
Danger
Synonyms: 5-Chloro-2-methylaniline, 2-Amino-4-chlorotoluene
OR17697-100G OR17697-500G OR17697-25G
APOSOR17697-100GEA 31.9 EUR
APOSOR17697-100G APOSOR17697-500G APOSOR17697-25G
5-Chloro-o-toluidine 98%
5-Chloro-o-toluidine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 236…238 °C
Melting Pt: 20…22 °C
Density: 1.18 g/cm³ (25 °C)
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00007779
CAS Number: 95-79-4
UN: 3429
ADR: 6.1 III