Quinoxaline-6-carbaldehyde
Supplier: Apollo Scientific
Warning
OR17708-100MG OR17708-250MG
APOSOR17708-100MGEA 21.9 EUR
APOSOR17708-100MG APOSOR17708-250MG
Quinoxaline-6-carbaldehyde
Quinoxaline-6-carbaldehyde
Formula: C9H6N2O
MW: 158.1600 g/mol
Melting Pt: 130…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00805834
CAS Number: 130345-50-5