4,4,5,5-Tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4,4,5,5-TETRAMETHYL-2-(1-METHYLETHENYL)-1,3,2-DIOXABOROLANE, 2-Isopropenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, (4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)isopropene
OR17811-100G OR17811-25G OR17811-5G
APOSOR17811-100GEA 132 EUR
APOSOR17811-100G APOSOR17811-25G APOSOR17811-5G
4,4,5,5-Tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(prop-1-en-2-yl)-1,3,2-dioxaborolane
Formula: C₉H₁₇BO₂
MW: 168.04 g/mol
Boiling Pt: 47…49 °C/6.75mmHg
Density: 0.894 g/cm³ (25 °C)
Flash Pt: 42 °C
Storage Temperature: Freezer
MDL Number: MFCD08276843
CAS Number: 126726-62-3
UN: 1993
ADR: 3,III