3,4-Ethylenedioxythiophene 98%
Supplier: Apollo Scientific
Danger
Synonyms: 2,3-Dihydrothieno[3,4-b][1,4]dioxine, EDOT
OR17830-100G OR17830-25G OR17830-5G
APOSOR17830-100GEA 183 EUR
APOSOR17830-100G APOSOR17830-25G APOSOR17830-5G
3,4-Ethylenedioxythiophene 98%
3,4-Ethylenedioxythiophene
Formula: C₆H₆O₂S
MW: 142.17 g/mol
Boiling Pt: 193 °C
Melting Pt: 10.5 °C
Density: 1.331 g/cm³ (25 °C)
Flash Pt: 104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093622
CAS Number: 126213-50-1
UN: 2810
ADR: 6.1 III