2-Bromoquinoline 97%
Supplier: Apollo Scientific
Warning
OR17842-1G OR17842-25G OR17842-5G
APOSOR17842-1GEA 21.9 EUR
APOSOR17842-1G APOSOR17842-25G APOSOR17842-5G
2-Bromoquinoline 97%
2-Bromoquinoline
Formula: C₉H₆BrN
MW: 208.06 g/mol
Melting Pt: 47…48 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00234480
CAS Number: 2005-43-8