4-Chloro-2,6-dimethylpyridine 98%
Supplier: Apollo Scientific
Warning
OR17851-1G OR17851-25G OR17851-5G
APOSOR17851-1GEA 26.1 EUR
APOSOR17851-1G APOSOR17851-25G APOSOR17851-5G
4-Chloro-2,6-dimethylpyridine 98%
4-Chloro-2,6-dimethylpyridine
Formula: C₇H₈ClN
MW: 141.6000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01646174
CAS Number: 3512-75-2