1-Methyl-1H-pyrazole-4-carboxylic acid
Supplier: Apollo Scientific
Warning
OR17854-1G OR17854-5G
APOSOR17854-1GEA 21.9 EUR
APOSOR17854-1G APOSOR17854-5G
1-Methyl-1H-pyrazole-4-carboxylic acid
1-Methyl-1H-pyrazole-4-carboxylic acid
Formula: C₅H₆N₂O₂
MW: 126.1100 g/mol
Melting Pt: 203…208 °C
Storage Temperature: Ambient
MDL Number: MFCD00159641
CAS Number: 5952-92-1