4-Chloro-o-toluidine
Supplier: Apollo Scientific
Danger
OR17861-100G OR17861-250G OR17861-25G
APOSOR17861-100GEA 52.2 EUR
APOSOR17861-100G APOSOR17861-250G APOSOR17861-25G
4-Chloro-o-toluidine
4-Chloro-o-toluidine
Formula: C₇H₈ClN
MW: 141.6 g/mol
Boiling Pt: 241 °C
Melting Pt: 24…30 °C
Density: 1.19 g/cm³ (25 °C)
Flash Pt: 124 °C
Storage Temperature: Ambient
MDL Number: MFCD00007842
CAS Number: 95-69-2
EINECS: 202-441-6
UN: 2239
ADR: 6.1,III