Bis(tri-tert-butylphosphine)palladium(0)
Supplier: Apollo Scientific
Warning
OR17884-5G OR17884-1G OR17884-250MG
APOSOR17884-5GEA 255 EUR
APOSOR17884-5G APOSOR17884-1G APOSOR17884-250MG
Bis(tri-tert-butylphosphine)palladium(0)
Bis(tri-tert-butylphosphine)palladium(0)
Formula: C₂₄H₅₄P₂Pd
MW: 511.05 g/mol
Melting Pt: 258…272 °C
Storage Temperature: Freezer
MDL Number: MFCD03094580
CAS Number: 53199-31-8