2,3-Dibromo-5-chloropyridine 98%
Supplier: Apollo Scientific
Warning
OR17894-100G OR17894-500G
APOSOR17894-100GEA 75.4 EUR
APOSOR17894-100G APOSOR17894-500G
2,3-Dibromo-5-chloropyridine 98%
2,3-Dibromo-5-chloropyridine
Formula: C₅H₂Br₂ClN
MW: 271.3400 g/mol
Melting Pt: 39…43 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08687254
CAS Number: 137628-17-2