Maleic acid
Supplier: Apollo Scientific
Warning
OR17920-100G OR17920-25G OR17920-10G
APOSOR17920-100GEA 109 EUR
APOSOR17920-100G APOSOR17920-25G APOSOR17920-10G
Maleic acid
Maleic acid
Formula: C₄H₄O₄
Boiling Pt: 160 °C
Melting Pt: 132…140 °C
Density: 1.59 g/cm³ (25 °C)
Flash Pt: 127 °C
Storage Temperature: Ambient
MDL Number: MFCD00063177
CAS Number: 110-16-7
EINECS: 203-742-5