3-Bromo-1H-pyrazolo[4,3-b]pyridine 98%
Supplier: Apollo Scientific
Warning
OR17930-1G OR17930-5G OR17930-10G
APOSOR17930-1GEA 78.3 EUR
APOSOR17930-1G APOSOR17930-5G APOSOR17930-10G
3-Bromo-1H-pyrazolo[4,3-b]pyridine 98%
3-Bromo-1H-pyrazolo[4,3-b]pyridine
Formula: C₆H₄BrN₃
MW: 198.0201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11518974
CAS Number: 633328-33-3