2-Bromo-m-toluidine 98%
Supplier: Apollo Scientific
Warning
OR17936-1G OR17936-5G
APOSOR17936-1GEA 21.9 EUR
APOSOR17936-1G APOSOR17936-5G
2-Bromo-m-toluidine 98%
2-Bromo-m-toluidine
Formula: C₇H₈BrN
Boiling Pt: 48 °C/0.03mmHg
Density: 1.489 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06411367
CAS Number: 54879-20-8