Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-aminothieno[2,3-b]pyridine-2-carboxylate
OR17991-10G OR17991-1G OR17991-5G
APOSOR17991-10GEA 203 EUR
APOSOR17991-10G APOSOR17991-1G APOSOR17991-5G
Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Methyl-3-aminothieno[2,3-b]pyridine-2-carboxylate
Formula: C₉H₈N₂O₂S
MW: 208.24 g/mol
Melting Pt: 198…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00174679
CAS Number: 111042-89-8