2-Bromo-3-nitrotoluene 95%
Supplier: Apollo Scientific
Warning
OR18002-100G OR18002-25G OR18002-5G
APOSOR18002-100GEA 157 EUR
APOSOR18002-100G APOSOR18002-25G APOSOR18002-5G
2-Bromo-3-nitrotoluene 95%
2-Bromo-3-nitrotoluene
Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
Boiling Pt: 135…136 °C/8mmHg
Melting Pt: 38…41 °C
Density: 0.803 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00134555
CAS Number: 41085-43-2