3-Bromo-6-methyl-5-nitro-1H-indazole
Supplier: Apollo Scientific
Warning
OR18056-1G OR18056-250MG
APOSOR18056-1GEA 24.8 EUR
APOSOR18056-1G APOSOR18056-250MG
3-Bromo-6-methyl-5-nitro-1H-indazole
3-Bromo-6-methyl-5-nitro-1H-indazole
Formula: C₈H₆BrN₃O₂
MW: 256.0561 g/mol
Melting Pt: 223…225 °C
Storage Temperature: Ambient
MDL Number: MFCD09026989
CAS Number: 1000343-58-7